missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diphenyl-1,6-hexanedione 98.0+%, TCI America™

Catalog No. d277325g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D277325G 25 g
D27735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D277325G Supplier TCI America Supplier No. D277325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3375-38-0
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00004761
InChI Key VRBLNWVVFVBNRK-UHFFFAOYSA-N
Synonym 1,4-Dibenzoylbutane
PubChem CID 76903
IUPAC Name 1,6-diphenylhexane-1,6-dione
SMILES C1=CC=C(C=C1)C(=O)CCCCC(=O)C2=CC=CC=C2

Specifications

Melting Point 107°C
Color White
Quantity 25 g
Formula Weight 266.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Diphenyl-1,6-hexanedione