accessibility menu, dialog, popup

UserName

CAS RN 3375-38-0

IUPAC Name: 1,6-diphenylhexane-1,6-dione
Synonyms: 1,6-diphenylhexane-1,6-dione
Molecular Weight (g/mol): 266.34
Molecular Formula: C18H18O2
InChi Key: VRBLNWVVFVBNRK-UHFFFAOYSA-N
SMILES: O=C(CCCCC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results