missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dihydroxynaphthalene 99.0+%, TCI America™

Catalog No. p-7126306
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059125G 25 g
D0591500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D059125G Supplier TCI America Supplier No. D059125G

Chemical Identifiers

CAS 575-44-0
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00003981
InChI Key FZZQNEVOYIYFPF-UHFFFAOYSA-N
Synonym 1,6-dihydroxynaphthalene, 1,6-naphthalenediol, 6-hydroxy-1-naphthol, 1,6-dihydroxy naphthalene, 2,5-naphthalenediol, 2,5-dihydroxynaphthalene, naphthalene, 1,6-dihydroxy, ccris 7894, 1,6-dihydroxynaphtalene, 1.6-dihydroxynaphthalene
PubChem CID 68463
ChEBI CHEBI:42040
IUPAC Name naphthalene-1,6-diol
SMILES C1=CC2=C(C=CC(=C2)O)C(=C1)O

Specifications

Melting Point 135°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Dihydroxynaphthalene
EINECSNumber (4)-0356
TSCA Yes