missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dichlorohexane 98.0+%, TCI America™

Catalog No. p-7126171
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D037225G 25 g
D0372500G 500 g
2 options

Catalog No. D037225G

Supplier: TCI America D037225G

Only null left

Chemical Identifiers

CAS 2163-00-0
Molecular Formula C6H12Cl2
Molecular Weight (g/mol) 155.062
MDL Number MFCD00001020
InChI Key OVISMSJCKCDOPU-UHFFFAOYSA-N
PubChem CID 16551
IUPAC Name 1,6-dichlorohexane
SMILES C(CCCCl)CCCl

Specifications

Color Colorless
Boiling Point 95°C
Quantity 25 g
UN Number 3082
Synonym hexane, 1,6-dichloro, hexamethylene dichloride, hexamethylene chloride, 1,6-dichlorhexan, hexane,6-dichloro, 1,6-dichloro-hexan, 1,6-dichloro hexane, hexane,1,6-dichloro, acmc-209fml
Formula Weight 155.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Dichlorohexane
EINECSNumber (2)-0064&(2)-0066
TSCA Yes