accessibility menu, dialog, popup

UserName

CAS RN 2163-00-0

IUPAC Name: 1,6-dichlorohexane
Synonyms: hexane, 1,6-dichloro-
Molecular Weight (g/mol): 155.06
Molecular Formula: C6H12Cl2
InChi Key: OVISMSJCKCDOPU-UHFFFAOYSA-N
SMILES: ClCCCCCCCl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)