missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dichlorohexane, 98%

Catalog No. AAB2176336
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2176336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2176336 Supplier Thermo Scientific Chemicals Supplier No. B2176336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2163-00-0 | C6H12Cl2 | 155.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2163-00-0
Molecular Formula C6H12Cl2
Molecular Weight (g/mol) 155.062
MDL Number MFCD00001020
InChI Key OVISMSJCKCDOPU-UHFFFAOYSA-N
Synonym hexane, 1,6-dichloro, hexamethylene dichloride, hexamethylene chloride, 1,6-dichlorhexan, hexane,6-dichloro, 1,6-dichloro-hexan, 1,6-dichloro hexane, hexane,1,6-dichloro, acmc-209fml
PubChem CID 16551
IUPAC Name 1,6-dichlorohexane
SMILES C(CCCCl)CCCl

Specifications

Melting Point -13°C
Density 1.067
Boiling Point 204°C to 205°C
Flash Point 73°C (163°F)
Refractive Index 1.4568
Quantity 500 g
UN Number UN3082
Beilstein 773673
Formula Weight 155.07
Percent Purity 98%
Chemical Name or Material 1,6-Dichlorohexane
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 218-491-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only