missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromohexane 97.0+%, TCI America™

Catalog No. D018525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D018525G 25 g
D0185100G 100 g
D0185500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D018525G Supplier TCI America Supplier No. D018525G

Chemical Identifiers

CAS 629-03-8
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00000272
InChI Key SGRHVVLXEBNBDV-UHFFFAOYSA-N
Synonym hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane
PubChem CID 12368
IUPAC Name 1,6-dibromohexane
SMILES C(CCCBr)CCBr

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 246°C
Quantity 25 g
Formula Weight 243.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Dibromohexane
RTECSNumber MO1515000
TSCA Yes