missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromohexane, 98%

Catalog No. AC112811000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112811000 100 mL Glass bottle
AC112815000 500 mL Glass bottle
AC112810025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112811000 Supplier Thermo Scientific Chemicals Supplier No. 112811000

CAS: 629-03-8 | C6H12Br2 | 243.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-03-8
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00000272
InChI Key SGRHVVLXEBNBDV-UHFFFAOYSA-N
Synonym hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane
PubChem CID 12368
IUPAC Name 1,6-dibromohexane
SMILES C(CCCBr)CCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -2.0°C
Color Colorless to Yellow
Density 1.5800g/mL
Boiling Point 243.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5056 to 1.5076
Linear Formula Br(CH2)6Br
Quantity 100 mL
Beilstein 01,145
Specific Gravity 1.58
Solubility Information Solubility in water: insoluble.
Formula Weight 243.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,6-Dibromohexane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation or rash occurs: Get medic
EINECSNumber 211-067-2
RTECSNumber MO1515000
TSCA TSCA

RUO – Research Use Only