missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromododecafluorohexane 98.0+%, TCI America™

Catalog No. p-7127797
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35721G 1 g
D35725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35721G Supplier TCI America Supplier No. D35721G

Chemical Identifiers

CAS 918-22-9
Molecular Formula C6Br2F12
Molecular Weight (g/mol) 459.86
MDL Number MFCD00153110
InChI Key SWAPKQWKFYIOJS-UHFFFAOYSA-N
Synonym 1,6-dibromododecafluorohexane, 1,6-dibromoperfluorohexane, hexane, 1,6-dibromo-dodecafluoro, dibromoperfluorohexane, acmc-209rdr, perfluoro 1,6-dibromohexane, hexane,1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro, 1,6-bis bromanyl-1,1,2,2,3,3,4,4,5,5,6,6-dodecakis fluoranyl hexane
PubChem CID 522992
IUPAC Name 1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexane
SMILES FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 459.85
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Dibromododecafluorohexane
TSCA No
Recommended Storage Refrigerator