missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Bis(acryloyloxy)hexane (stabilized with MEHQ) 85.0+%, TCI America™
Supplier: TCI America B293625G
Specifications
| 1,6-Bis(acryloyloxy)hexane (stabilized with MEHQ) | |
| Yellow | |
| C12H18O4 | |
| 25 g | |
| 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate | |
| C=CC(=O)OCCCCCCOC(=O)C=C | |
| 226.27 | |
| 226.27 | |
| Liquid |
| 13048-33-4 | |
| 165°C | |
| MFCD00008631 | |
| 3082 | |
| FIHBHSQYSYVZQE-UHFFFAOYSA-N | |
| 6-(prop-2-enoyloxy)hexyl prop-2-enoate | |
| 25644 | |
| ≥85.0% (GC) |
Chemical Identifiers
| 13048-33-4 | |
| 226.27 | |
| FIHBHSQYSYVZQE-UHFFFAOYSA-N | |
| 25644 | |
| C=CC(=O)OCCCCCCOC(=O)C=C |
| C12H18O4 | |
| MFCD00008631 | |
| 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate | |
| 6-(prop-2-enoyloxy)hexyl prop-2-enoate |
Safety and Handling
EINECSNumber : (2)-1007&(2)-0958
RTECSNumber : AT1430000
TSCA : Yes