missing translation for 'onlineSavingsMsg'
Learn More

1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane (stabilized with MEHQ) 90.0+%, TCI America™

Catalog No. B52781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52781G 1 g
B52785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52781G Supplier TCI America Supplier No. B52781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2264-01-9
Molecular Formula C12H10F8O4
Molecular Weight (g/mol) 370.20
MDL Number MFCD00078412
InChI Key BWTMTZBMAGYMOD-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-Octafluorohexamethylene Diacrylate, 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol Diacrylate
PubChem CID 2775863
IUPAC Name (2,2,3,3,4,4,5,5-octafluoro-6-prop-2-enoyloxyhexyl) prop-2-enoate
SMILES C=CC(=O)OCC(C(C(C(COC(=O)C=C)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 370.20
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane (stabilized with MEHQ)