accessibility menu, dialog, popup

UserName

H2COOOOCH2FFFFFFFF

CAS RN 2264-01-9

IUPAC Name: 2,2,3,3,4,4,5,5-octafluoro-6-(prop-2-enoyloxy)hexyl prop-2-enoate
Synonyms: 2,2,3,3,4,4,5,5-octafluoro-6-(prop-2-enoyloxy)hexyl prop-2-enoate
Molecular Weight (g/mol): 370.2
Molecular Formula: C12H10F8O4
InChi Key: BWTMTZBMAGYMOD-UHFFFAOYSA-N
SMILES: FC(F)(COC(=O)C=C)C(F)(F)C(F)(F)C(F)(F)COC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results