missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,6-Anhydro-beta-D-glucopyranose, 99+%

Catalog No. p-3756863
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270950010 1 g Glass bottle
AC270950050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270950010 Supplier Thermo Scientific Chemicals Supplier No. 270950010
Only null left

CAS: 498-07-7 | C6H10O5 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 498-07-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00063248
InChI Key TWNIBLMWSKIRAT-UHFFFAOYNA-N
Synonym 1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucose
PubChem CID 2724705
ChEBI CHEBI:30997
SMILES OC1C2COC(O2)C(O)C1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 170.0°C to 180.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation − 66.00
Solubility Information (5% in water) Clear colorless
IUPAC Name (1R,2S,3S,4R,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
Formula Weight 162.14
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1,6-Anhydro-β-D-glucopyranose
Show More Show Less
EINECSNumber 207-855-

RUO – Research Use Only