missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentanediol 97.0+%, TCI America™

Catalog No. P005025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P005025G 25 g
P0050500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P005025G Supplier TCI America Supplier No. P005025G

Chemical Identifiers

CAS 111-29-5
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002978
InChI Key ALQSHHUCVQOPAS-UHFFFAOYSA-N
Synonym 1,5-pentanediol, 1,5-dihydroxypentane, pentylene glycol, pentamethylene glycol, 1,5-pentylene glycol, 1,5 pentanediol, 1,5-pentamethylene glycol, alpha,omega-pentanediol, unii-07uxz0scst, 1,5-pentandiol
PubChem CID 8105
IUPAC Name pentane-1,5-diol
SMILES OCCCCCO

Specifications

Melting Point -16°C
Color Colorless
Boiling Point 239°C
Quantity 25 g
Formula Weight 104.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Pentanediol
EINECSNumber (2)-0240
RTECSNumber SA0480000
TSCA Yes