missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentamethylenetetrazole 98.0+%, TCI America™

Supplier:  TCI America P004625G

 View more versions of this product

Catalog No. P004625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,5-Pentamethylenetetrazole
60°C
194°C
MFCD00005939
2811
CWRVKFFCRWGWCS-UHFFFAOYSA-N
6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
5917
138.17
Crystalline Powder
54-95-5
White-Yellow
C6H10N4
25 g
pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
C1CCC2=NN=NN2CC1
138.174
CHEBI:34910
≥98.0% (GC)

Chemical Identifiers

54-95-5
138.174
CWRVKFFCRWGWCS-UHFFFAOYSA-N
5917
6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
C6H10N4
MFCD00005939
pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
CHEBI:34910
C1CCC2=NN=NN2CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XF8225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions