missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentamethylenetetrazole 98.0+%, TCI America™

Catalog No. p-7134390
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00465G 5 g
P004625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00465G Supplier TCI America Supplier No. P00465G
Only null left

Chemical Identifiers

CAS 54-95-5
Molecular Formula C6H10N4
Molecular Weight (g/mol) 138.174
MDL Number MFCD00005939
InChI Key CWRVKFFCRWGWCS-UHFFFAOYSA-N
Synonym pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
PubChem CID 5917
ChEBI CHEBI:34910
IUPAC Name 6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
SMILES C1CCC2=NN=NN2CC1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 194°C
Quantity 5 g
UN Number 2811
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Pentamethylenetetrazole
RTECSNumber XF8225000
TSCA Yes