missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine 98.0+%, TCI America™

Catalog No. p-7129809
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0650100MG 100 mg
F06501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0650100MG Supplier TCI America Supplier No. F0650100MG

Chemical Identifiers

CAS 928830-73-3
Molecular Formula C11H13FN4O4
Molecular Weight (g/mol) 284.25
MDL Number MFCD10000959
InChI Key SSBAFRIGYXRBRK-UHFFFAOYSA-N
Synonym PPZ
PubChem CID 16223480
IUPAC Name 1-(5-fluoro-2,4-dinitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 115°C
Color Yellow
Quantity 100 mg
Formula Weight 284.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine
TSCA No
Recommended Storage Refrigerator