missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbonohydrazide 98.0+%, TCI America™

Catalog No. D151325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D151325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D151325G Supplier TCI America Supplier No. D151325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00003013
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 175°C
Quantity 25 g
Formula Weight 242.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diphenylcarbonohydrazide
EINECSNumber (3)-2202
RTECSNumber FF2750000
TSCA Yes
Recommended Storage Freezer