missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diphenylcarbohydrazide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$3445.12 - $3445.12
Chemical Identifiers
| CAS | 140-22-7 |
|---|---|
| Molecular Formula | C13H14N4O |
| Molecular Weight (g/mol) | 242.282 |
| InChI Key | KSPIHGBHKVISFI-UHFFFAOYSA-N |
| Synonym | 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl |
| PubChem CID | 8789 |
| ChEBI | CHEBI:4641 |
| IUPAC Name | 1,3-dianilinourea |
| SMILES | C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
Chemical Identifiers
| 140-22-7 | |
| 242.282 | |
| 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
| CHEBI:4641 | |
| C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
| C13H14N4O | |
| KSPIHGBHKVISFI-UHFFFAOYSA-N | |
| 8789 | |
| 1,3-dianilinourea |
Specifications
| 140-22-7 | |
| C13H14N4O | |
| 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
| C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 | |
| 242.282 | |
| CHEBI:4641 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1,5-Diphenylcarbohydrazide |
| Wide Mouth Amber Glass Bottle | |
| 100 g | |
| KSPIHGBHKVISFI-UHFFFAOYSA-N | |
| 1,3-dianilinourea | |
| 8789 | |
| 242.28 | |
| Powder |