missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbazide, ≥98% (HPLC), Reag. Ph. Eur., MilliporeSigma™ Supelco™

Catalog No. 111013050
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013050 10 g
111013051 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013050 Supplier Merck Emd Millipore Supplier No. 3315210G

1,5-Diphenylcarbazide reaction with chromium(VI), has been widely used for the spectro- photometric determination of chromium. It is also an electron donor, and can be used for measuring photosystem 2 activity in subchloroplast fragments.

Chemical Identifiers

MDL Number MFCD00003013
Synonym 1,5-Diphenylcarbohydrazide; sym.-Diphenylcarbazide

Specifications

Melting Point 170°C to 175°C; 170°C to 175°C
CAS 140-22-7
Linear Formula C6H5 NHNHCONHNHC6H5
Quantity 10 g
Formula Weight 242.28
Percent Purity ≥98% (HPLC); ≥98.0%
Grade Reag. Ph. Eur.
RTECSNumber FF2750000