missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbazide, 97+%

Catalog No. AAA1406036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1406036 500 g
AAA1406014 25 g
AAA1406022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1406036 Supplier Thermo Scientific Chemicals Supplier No. A1406036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 140-22-7 | C13H14N4O | 242.282 g/mol

1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

Solubility
Soluble in glacial acetic acid, alcohol and acetone. Slightly soluble in water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00003013
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 170°C to 176°C
Quantity 500 g
Beilstein 752039
Sensitivity Air and light sensitive
Merck Index 14,3321
Solubility Information Soluble in glacial acetic acid,alcohol and acetone. Slightly soluble in water.
Formula Weight 242.28
Percent Purity ≥97%
Chemical Name or Material 1,5-Diphenylcarbazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-403-7
RTECSNumber FF2750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only