missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dioxepan-2-one 98.0+%, TCI America™

Catalog No. p-7127875
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37075G 5 g
D370725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37075G Supplier TCI America Supplier No. D37075G

Chemical Identifiers

CAS 35438-57-4
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
InChI Key AOLNDUQWRUPYGE-UHFFFAOYSA-N
PubChem CID 193214
IUPAC Name 1,4-dioxepan-5-one
SMILES C1COCCOC1=O

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 5 g
Formula Weight 116.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dioxepan-2-one
TSCA No
Recommended Storage Freezer