accessibility menu, dialog, popup

UserName

OOO

CAS RN 35438-57-4

IUPAC Name: 1,4-dioxepan-5-one
Synonyms: 1,4-dioxepan-5-one
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: AOLNDUQWRUPYGE-UHFFFAOYSA-N
SMILES: O=C1CCOCCO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results