missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 95+%, Thermo Scientific™

Catalog No. CC12601EA
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12601EA 10g
CC12601DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12601EA Supplier Thermo Scientific Chemicals Supplier No. CC12601EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5744-59-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00085040
InChI Key PXRXGHUTKHXUGF-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 587757
IUPAC Name 1,5-dimethylpyrazole-3-carboxylic acid
SMILES CN1N=C(C=C1C)C(O)=O

Specifications

Assay Percent Range 95%
Quantity 10g
Formula Weight 140.14
Percent Purity 95%
Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid
Hazard Category Warning