missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC12602DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12602DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12602DA Supplier Thermo Scientific Chemicals Supplier No. CC12602DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 49783-84-8
Molecular Formula C6H7ClN2O
Molecular Weight (g/mol) 158.59
MDL Number MFCD03407950
InChI Key BADBQYAILRGCBB-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrazole-3-carbonyl chloride, 1h-pyrazole-3-carbonylchloride, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carbonylchloride, 1h-pyrazole-3-carbonyl chloride, 1,5-dimethyl-9ci
PubChem CID 2776364
IUPAC Name 1,5-dimethylpyrazole-3-carbonyl chloride
SMILES CN1N=C(C=C1C)C(Cl)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 158.59
Percent Purity 97%
Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.