CAS RN 49783-84-8
IUPAC Name:
1,5-dimethyl-1H-pyrazole-3-carbonyl chloride
Synonyms:
1,5-dimethylpyrazole-3-carbonyl chloride
Molecular Weight (g/mol):
158.59
Molecular Formula:
C6H7ClN2O
InChi Key:
BADBQYAILRGCBB-UHFFFAOYSA-N
SMILES:
CN1N=C(C=C1C)C(Cl)=O
1
–
1
of
1
results
1
1,5-Dimethyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™
CAS: 49783-84-8 Molecular Formula: C6H7ClN2O Molecular Weight (g/mol): 158.59 MDL Number: MFCD03407950 InChI Key: BADBQYAILRGCBB-UHFFFAOYSA-N Synonym: 1,5-dimethyl-1h-pyrazole-3-carbonyl chloride,1h-pyrazole-3-carbonylchloride, 1,5-dimethyl,1,5-dimethyl-1h-pyrazole-3-carbonylchloride,1h-pyrazole-3-carbonyl chloride, 1,5-dimethyl-9ci PubChem CID: 2776364 IUPAC Name: 1,5-dimethylpyrazole-3-carbonyl chloride SMILES: CN1N=C(C=C1C)C(Cl)=O