missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethoxypentane 97.0+%, TCI America™

Catalog No. D28575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28575G Supplier TCI America Supplier No. D28575G

Chemical Identifiers

CAS 111-89-7
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.20
MDL Number MFCD03613608
InChI Key NFCBHIFOKVDBCJ-UHFFFAOYSA-N
Synonym Pentamethylene Glycol Dimethyl Ether, 1,5-Pentanediol Dimethyl Ether
PubChem CID 8145
IUPAC Name 1,5-dimethoxypentane
SMILES COCCCCCOC

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 132.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Dimethoxypentane