accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 111-89-7

IUPAC Name: 1,5-dimethoxypentane
Synonyms: 1,5-dimethoxypentane
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: NFCBHIFOKVDBCJ-UHFFFAOYSA-N
SMILES: COCCCCCOC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results