missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dimercaptonaphthalene 98.0+%, TCI America™
$281.48 - $846.29
Chemical Identifiers
| CAS | 5325-88-2 |
|---|---|
| Molecular Formula | C10H8S2 |
| Molecular Weight (g/mol) | 192.294 |
| MDL Number | MFCD00068909 |
| InChI Key | AAPAGLBSROJFGM-UHFFFAOYSA-N |
| Synonym | 1,5-Naphthalenedithiol |
| PubChem CID | 21397 |
| IUPAC Name | naphthalene-1,5-dithiol |
| SMILES | C1=CC2=C(C=CC=C2S)C(=C1)S |
Chemical Identifiers
| 5325-88-2 | |
| 192.294 | |
| AAPAGLBSROJFGM-UHFFFAOYSA-N | |
| 21397 | |
| C1=CC2=C(C=CC=C2S)C(=C1)S |
| C10H8S2 | |
| MFCD00068909 | |
| 1,5-Naphthalenedithiol | |
| naphthalene-1,5-dithiol |
Specifications
| 5325-88-2 | |
| White-Yellow | |
| MFCD00068909 | |
| 1,5-Naphthalenedithiol | |
| C1=CC2=C(C=CC=C2S)C(=C1)S | |
| 192.294 | |
| 192.29 | |
| Crystalline Powder |
| 121°C | |
| C10H8S2 | |
| 1 g | |
| AAPAGLBSROJFGM-UHFFFAOYSA-N | |
| naphthalene-1,5-dithiol | |
| 21397 | |
| ≥98.0% (GC,T) | |
| 1,5-Dimercaptonaphthalene |
Safety and Handling
TSCA : No