missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diisocyanatonaphthalene 98.0+%, TCI America™

$273.38 - $668.03

Chemical Identifiers

CAS 3173-72-6
Molecular Formula C12H6N2O2
Molecular Weight (g/mol) 210.192
InChI Key SBJCUZQNHOLYMD-UHFFFAOYSA-N
Synonym Naphthalene 1,5-Diisocyanate
PubChem CID 18503
ChEBI CHEBI:82496
IUPAC Name 1,5-diisocyanatonaphthalene
SMILES C1=CC2=C(C=CC=C2N=C=O)C(=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
N016825G
View Documents
TCI America
N016825G
25 g
Each for $273.38
Only null left
 
N0168100G
View Documents
TCI America
N0168100G
100 g
Each for $668.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3173-72-6
210.192
Naphthalene 1,5-Diisocyanate
CHEBI:82496
C1=CC2=C(C=CC=C2N=C=O)C(=C1)N=C=O
C12H6N2O2
SBJCUZQNHOLYMD-UHFFFAOYSA-N
18503
1,5-diisocyanatonaphthalene

Specifications

3173-72-6
White-Yellow
C12H6N2O2
2206
SBJCUZQNHOLYMD-UHFFFAOYSA-N
1,5-diisocyanatonaphthalene
18503
210.19
Crystalline Powder
129°C
183°C
25 g
Naphthalene 1,5-Diisocyanate
C1=CC2=C(C=CC=C2N=C=O)C(=C1)N=C=O
210.192
CHEBI:82496
≥98.0% (GC)
1,5-Diisocyanatonaphthalene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0527

RTECSNumber : NQ9600000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents