accessibility menu, dialog, popup

UserName

CAS RN 3173-72-6

OCNNCO

CAS RN 3173-72-6

IUPAC Name: 1,5-diisocyanatonaphthalene
Synonyms: 1,5-naphthylene di-isocyanate
Molecular Weight (g/mol): 210.19
Molecular Formula: C12H6N2O2
InChi Key: SBJCUZQNHOLYMD-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC2=C(C=CC=C12)N=C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results