missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diiodopentane, 97%, stab. with copper

Catalog No. p-7023588
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1729814 25 g
AAA1729822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1729814 Supplier Thermo Scientific Chemicals Supplier No. A1729814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-77-3 | C5H10I2 | 323.944 g/mol

1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 628-77-3
Molecular Formula C5H10I2
Molecular Weight (g/mol) 323.944
MDL Number MFCD00001101
InChI Key IAEOYUUPFYJXHN-UHFFFAOYSA-N
Synonym pentane, 1,5-diiodo, pentamethylene diiodide, 1,5-diiodopentan, pentane,5-diiodo, pubchem3853, acmc-20ap1i, 4-01-00-00317 beilstein handbook reference, iaeoyuupfyjxhn-uhfffaoysa, 1,5-diiodopentane, stab. with copper, 1,5-diiodopentane, contains copper as stabilizer
PubChem CID 12354
IUPAC Name 1,5-diiodopentane
SMILES C(CCI)CCI

Specifications

Density 2.176
Boiling Point 128°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6012
Quantity 25 g
Beilstein 1697281
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 323.94
Percent Purity 97%
Chemical Name or Material 1,5-Diiodopentane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-054-1
RTECSNumber SA0425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only