accessibility menu, dialog, popup

UserName

CAS RN 628-77-3

IUPAC Name: 1,5-diiodopentane
Synonyms: pentane, 1,5-diiodo-
Molecular Weight (g/mol): 323.94
Molecular Formula: C5H10I2
InChi Key: IAEOYUUPFYJXHN-UHFFFAOYSA-N
SMILES: ICCCCCI
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)