missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dihydroxynaphthalene 98.0+%, TCI America™
$61.51 - $490.25
Chemical Identifiers
| CAS | 83-56-7 |
|---|---|
| Molecular Formula | C10H8O2 |
| Molecular Weight (g/mol) | 160.172 |
| MDL Number | MFCD00003980 |
| InChI Key | BOKGTLAJQHTOKE-UHFFFAOYSA-N |
| Synonym | 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 |
| PubChem CID | 6749 |
| IUPAC Name | naphthalene-1,5-diol |
| SMILES | C1=CC2=C(C=CC=C2O)C(=C1)O |
Chemical Identifiers
| 83-56-7 | |
| 160.172 | |
| BOKGTLAJQHTOKE-UHFFFAOYSA-N | |
| 6749 | |
| C1=CC2=C(C=CC=C2O)C(=C1)O |
| C10H8O2 | |
| MFCD00003980 | |
| 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 | |
| naphthalene-1,5-diol |
Specifications
| 83-56-7 | |
| MFCD00003980 | |
| 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 | |
| C1=CC2=C(C=CC=C2O)C(=C1)O | |
| 160.172 | |
| 160.17 | |
| Crystalline Powder |
| C10H8O2 | |
| 25 g | |
| BOKGTLAJQHTOKE-UHFFFAOYSA-N | |
| naphthalene-1,5-diol | |
| 6749 | |
| ≥98.0% (GC) | |
| 1,5-Dihydroxynaphthalene |
Safety and Handling
EINECSNumber : (4)-0356
RTECSNumber : QJ4740000
TSCA : Yes