missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dihydroxynaphthalene 98.0+%, TCI America™

Catalog No. D059025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059025G 25 g
D0590100G 100 g
D0590500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D059025G Supplier TCI America Supplier No. D059025G

Chemical Identifiers

CAS 83-56-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00003980
InChI Key BOKGTLAJQHTOKE-UHFFFAOYSA-N
Synonym 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990
PubChem CID 6749
IUPAC Name naphthalene-1,5-diol
SMILES C1=CC2=C(C=CC=C2O)C(=C1)O

Specifications

Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dihydroxynaphthalene
EINECSNumber (4)-0356
RTECSNumber QJ4740000
TSCA Yes