Learn More
1,5-Dihydroxynaphthalene, 98%
CAS: 83-56-7 | C10H8O2 | 160.172 g/mol
$129.75 - $420.14
Chemical Identifiers
| CAS | 83-56-7 |
|---|---|
| Molecular Formula | C10H8O2 |
| Molecular Weight (g/mol) | 160.172 |
| MDL Number | MFCD00003980 |
| InChI Key | BOKGTLAJQHTOKE-UHFFFAOYSA-N |
| Synonym | 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 |
| PubChem CID | 6749 |
| IUPAC Name | naphthalene-1,5-diol |
| SMILES | C1=CC2=C(C=CC=C2O)C(=C1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0459930
|
Thermo Scientific Chemicals
L0459930 |
250 g |
Each for $129.75
|
|
|||||
|
AAL045990B
|
Thermo Scientific Chemicals
L045990B |
1000 g |
Each for $420.14
|
|
|||||
Description
1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 83-56-7 | |
| 160.172 | |
| BOKGTLAJQHTOKE-UHFFFAOYSA-N | |
| 6749 | |
| C1=CC2=C(C=CC=C2O)C(=C1)O |
| C10H8O2 | |
| MFCD00003980 | |
| 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 | |
| naphthalene-1,5-diol |
Specifications
| 83-56-7 | |
| 252°C (486°F) | |
| MFCD00003980 | |
| 2044951 | |
| Soluble in water. | |
| C1=CC2=C(C=CC=C2O)C(=C1)O | |
| 160.172 | |
| 160.17 | |
| 1,5-Dihydroxynaphthalene |
| ∽260°C (decomposition) | |
| C10H8O2 | |
| 250 g | |
| 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 | |
| BOKGTLAJQHTOKE-UHFFFAOYSA-N | |
| naphthalene-1,5-diol | |
| 6749 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
H302-H317-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 201-487-4
RTECSNumber : QJ4740000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only