Learn More
1,5-Dihydroxynaphthalene, 97%
CAS: 83-56-7 | C10H8O2 | 160.17 g/mol
$367.78 - $367.78
Chemical Identifiers
| CAS | 83-56-7 |
|---|---|
| Molecular Formula | C10H8O2 |
| Molecular Weight (g/mol) | 160.17 |
| InChI Key | BOKGTLAJQHTOKE-UHFFFAOYSA-N |
| Synonym | 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 |
| PubChem CID | 6749 |
| IUPAC Name | naphthalene-1,5-diol |
| SMILES | C1=CC2=C(C=CC=C2O)C(=C1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC115005000
|
Thermo Scientific Chemicals
115005000 |
500 g |
Each for $367.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83-56-7 | |
| 160.17 | |
| 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 | |
| naphthalene-1,5-diol |
| C10H8O2 | |
| BOKGTLAJQHTOKE-UHFFFAOYSA-N | |
| 6749 | |
| C1=CC2=C(C=CC=C2O)C(=C1)O |
Specifications
| 83-56-7 | |
| 252°C | |
| 96% min. (HPLC) | |
| C10H8O2 | |
| 500 g | |
| 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990 | |
| BOKGTLAJQHTOKE-UHFFFAOYSA-N | |
| naphthalene-1,5-diol | |
| 6749 | |
| 97% |
| 259.0°C to 261.0°C | |
| Authentic | |
| Plastic bottle | |
| C10H6(OH)2 | |
| 06,98 | |
| Solubility in water: 0.6g/L in water (20°C). Other solubilities: soluble in methanol | |
| C1=CC2=C(C=CC=C2O)C(=C1)O | |
| 160.17 | |
| 160.17 | |
| 1,5-Dihydroxynaphthalene |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
EINECSNumber : 201-487-4
RUO – Research Use Only