missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dihydroxynaphthalene, 97%

Catalog No. AC115005000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC115005000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC115005000 Supplier Thermo Scientific Chemicals Supplier No. 115005000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83-56-7 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83-56-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
InChI Key BOKGTLAJQHTOKE-UHFFFAOYSA-N
Synonym 1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990
PubChem CID 6749
IUPAC Name naphthalene-1,5-diol
SMILES C1=CC2=C(C=CC=C2O)C(=C1)O

Specifications

Melting Point 259.0°C to 261.0°C
Flash Point 252°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula C10H6(OH)2
Quantity 500 g
Beilstein 06,98
Solubility Information Solubility in water: 0.6g/L in water (20°C). Other solubilities: soluble in methanol
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 1,5-Dihydroxynaphthalene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
EINECSNumber 201-487-4

RUO – Research Use Only