missing translation for 'onlineSavingsMsg'
Learn More

1,5-Difluoro-2,4-dinitrobenzene 97.0+%, TCI America™

Catalog No. D16495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16495G 5 g
D164925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16495G Supplier TCI America Supplier No. D16495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 327-92-4
Molecular Formula C6H2F2N2O4
Molecular Weight (g/mol) 204.09
MDL Number MFCD00007052
InChI Key VILFTWLXLYIEMV-UHFFFAOYSA-N
Synonym 1,3-difluoro-4,6-dinitrobenzene, benzene, 1,5-difluoro-2,4-dinitro, dfdnb, 4,6-difluoro-1,3-dinitrobenzene, unii-g5vv4mq22v, ffdnb, g5vv4mq22v, 2,4-difluoro-1,5-dinitrobenzene, pubchem4398, acmc-1cli3
PubChem CID 67598
IUPAC Name 1,5-difluoro-2,4-dinitrobenzene
SMILES [O-][N+](=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O

Specifications

Melting Point 74°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 204.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Difluoro-2,4-dinitrobenzene
RTECSNumber CZ5663200
TSCA Yes