missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diethynyl-2,4-dimethylbenzene 98.0+%, TCI America™

Catalog No. p-7128520
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48781G 1 g
D48785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48781G Supplier TCI America Supplier No. D48781G
Only null left

Chemical Identifiers

CAS 1379822-09-9
Molecular Formula C12H10
Molecular Weight (g/mol) 154.212
InChI Key HPFZIHSKAUBEON-UHFFFAOYSA-N
PubChem CID 102363018
IUPAC Name 1,5-diethynyl-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1C#C)C#C)C

Specifications

Melting Point 58°C
Quantity 1 g
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diethynyl-2,4-dimethylbenzene