accessibility menu, dialog, popup

UserName

CAS RN 1379822-09-9

CH3HCCHCH3

CAS RN 1379822-09-9

IUPAC Name: 1,5-diethynyl-2,4-dimethylbenzene
Synonyms: 1,5-diethynyl-2,4-dimethylbenzene
Molecular Weight (g/mol): 154.21
Molecular Formula: C12H10
InChi Key: HPFZIHSKAUBEON-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1C#C)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results