missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromopentane 98.0+%, TCI America™

Catalog No. D019825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D019825G 25 g
D0198100G 100 g
D0198500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D019825G Supplier TCI America Supplier No. D019825G

Chemical Identifiers

CAS 111-24-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000268
InChI Key IBODDUNKEPPBKW-UHFFFAOYSA-N
PubChem CID 8100
IUPAC Name 1,5-dibromopentane
SMILES C(CCBr)CCBr

Specifications

Color Yellow
Boiling Point 221°C
Quantity 25 g
Synonym pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918
Formula Weight 229.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Dibromopentane
EINECSNumber (2)-3344
RTECSNumber SA0320000
TSCA Yes