missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromopentane, 98%

Catalog No. AAA1846536
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1846536 500 g
AAA1846522 100 g
AAA184650E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1846536 Supplier Thermo Scientific Chemicals Supplier No. A1846536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-24-0 | C5H10Br2 | 229.943 g/mol

1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

Solubility
Miscible with water, slightly miscible with chloroform and benzene.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 111-24-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000268
InChI Key IBODDUNKEPPBKW-UHFFFAOYSA-N
PubChem CID 8100
IUPAC Name 1,5-dibromopentane
SMILES C(CCBr)CCBr

Specifications

Melting Point -34°C
Density 1.688
Boiling Point 224°C
Flash Point 79°C (174°F)
Refractive Index 1.5122
Quantity 500 g
Beilstein 1209245
Synonym pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918
Solubility Information Miscible with water,slightly miscible with chloroform and benzene.
Formula Weight 229.95
Percent Purity 98%
Chemical Name or Material 1,5-Dibromopentane
Show More Show Less
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-849-7
RTECSNumber SA0320000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only