missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-3-methylpentane, 98+%

Catalog No. AAB2052106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2052106 5 g
AAB2052114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2052106 Supplier Thermo Scientific Chemicals Supplier No. B2052106

CAS: 4457-72-1 | C6H12Br2 | 243.97 g/mol

1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 4457-72-1
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00039200
InChI Key YDPZWUMQKMLLHC-UHFFFAOYSA-N
Synonym 3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
PubChem CID 138258
IUPAC Name 1,5-dibromo-3-methylpentane
SMILES CC(CCBr)CCBr

Specifications

Density 1.58
Boiling Point 233°C
Flash Point 101°C (214°F)
Refractive Index 1.508
Linear Formula BrCH2CH2CH(CH3)CH2CH2Br
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 243.98
Percent Purity ≥98%
Chemical Name or Material 1,5-Dibromo-3-methylpentane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only