missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminopentane 98.0+%, TCI America™

Catalog No. p-7126037
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01085ML 5 mL
D010825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01085ML Supplier TCI America Supplier No. D01085ML
Only null left

Chemical Identifiers

CAS 462-94-2
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008239
InChI Key VHRGRCVQAFMJIZ-UHFFFAOYSA-N
Synonym 1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p
PubChem CID 273
ChEBI CHEBI:18127
IUPAC Name pentane-1,5-diamine
SMILES NCCCCCN

Specifications

Boiling Point 180°C
Quantity 5 mL
UN Number 2735
Formula Weight 102.18
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1,5-Diaminopentane
EINECSNumber (2)-4146
RTECSNumber SA0200000
TSCA No