missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadiene 98.0+%, TCI America™

Catalog No. C050325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C050325ML 25 mL
C0503100ML 100 mL
C0503500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C050325ML Supplier TCI America Supplier No. C050325ML

Chemical Identifiers

CAS 111-78-4
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001752
InChI Key VYXHVRARDIDEHS-QGTKBVGQSA-N
Synonym 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
PubChem CID 10937607
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene
SMILES C1C\C=C/CC\C=C/1

Specifications

Melting Point -69°C
Color Yellow
Boiling Point 150°C
Quantity 25 mL
UN Number 2520
Formula Weight 108.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Cyclooctadiene [stabilized with Octadecyl 3-(3′,5′-Di-tert-butyl-4′-hydroxyphe
EINECSNumber (3)-2232
RTECSNumber GX9560000
TSCA Yes