missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) Hexafluorophosphate, TCI America™

Catalog No. C2824100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2824100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2824100MG Supplier TCI America Supplier No. C2824100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64536-78-3
Molecular Formula C31H50F6IrNP2
Molecular Weight (g/mol) 804.90
MDL Number MFCD00075097
InChI Key UJXHUUQZACSUOG-KJWGIZLLSA-N
Synonym crabtree's catalyst, unii-816rs2nbpn, 816rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate
PubChem CID 5702647
IUPAC Name iridium(1+) (1Z,5Z)-cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide pyridine tricyclohexylphosphane
SMILES [Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 175°C
Quantity 100 mg
Formula Weight 804.90
Physical Form Crystalline Powder
Chemical Name or Material (1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) Hexafluorophosphate