missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Cyclooctadiene)(methoxy)iridium(I) Dimer, TCI America™

Catalog No. C2662200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2662200MG 200 mg
C26621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2662200MG Supplier TCI America Supplier No. C2662200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12148-71-9
Molecular Formula C18H30Ir2O2
Molecular Weight (g/mol) 662.87
MDL Number MFCD08459360
InChI Key SGBSCFWMCJNCRM-MIXQCLKLSA-N
Synonym bis 1,5-cyclooctadiene di-mu-methoxydiiridium i, 1,5-cyclooctadiene methoxy iridium i dimer
PubChem CID 124202702
IUPAC Name bis((1Z,5Z)-cycloocta-1,5-diene); bis(methoxyiridiumylium)
SMILES CO[Ir+].CO[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 179°C
Color Yellow
Quantity 200 mg
Formula Weight 662.87
Physical Form Crystalline Powder
Chemical Name or Material (1,5-Cyclooctadiene)(methoxy)iridium(I) Dimer
TSCA No
Recommended Storage Refrigerator