accessibility menu, dialog, popup

UserName

CAS RN 12148-71-9

IUPAC Name: bis((1Z,5Z)-cycloocta-1,5-diene); bis(methoxyiridiumylium)
Synonyms: bis(1,5-cyclooctadiene, (Z,Z)-); bis(methoxyiridiumylium)
Molecular Weight (g/mol): 662.87
Molecular Formula: C18H30Ir2O2
InChi Key: SGBSCFWMCJNCRM-MIXQCLKLSA-N
SMILES: CO[Ir+].CO[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results