missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadiene, 99%, stabilized

CAS: 111-78-4 | C8H12 | 108.18 g/mol

Supplier:  Thermo Scientific Chemicals 297120025

 View more versions of this product

Catalog No. AC297120025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 5-Cyclooctadiene
111-78-4
100.0
Colorless
151°C (101.3 kPa)
Authentic
Glass bottle
1.4920 to 1.4940
2.5 L
0.88
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
VYXHVRARDIDEHS-QGTKBVGQSA-N
(1Z,5Z)-cycloocta-1,5-diene
50 to 150ppm t-butylcatechol
1.3 mPa.s (20°C)
99%
Stabilized
98.5
-69.5°C
0.8800g/mL
38°C
98.5% min. (GC)
C8H12
MFCD00001752
05, 116
15, 2730
Solubility in water: 780mg/l (20°C). Other solubilities: soluble in benzene and carbon tetrachloride
C1C\C=C/CC\C=C/1
108.18
10937607
108.18
Liquid

Chemical Identifiers

111-78-4
108.18
VYXHVRARDIDEHS-QGTKBVGQSA-N
10937607
C8H12
MFCD00001752
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
C1C\C=C/CC\C=C/1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger

EINECSNumber : 203-907-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only